missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ BRL 44408 maleate

Product Code. 30002138
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002138 10 mg 10mg
18065122 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002138 Supplier R&D Systems™ Supplier No. 1133/10
  • ONLINE EXCLUSIVE PRICE
    £193.30
    £173.25 / 10mg
    Save £20.05
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective α2A antagonist

BRL 44408 maleate is a selective α2A-adrenoceptor antagonist (Ki = 1.7 nM and 144.5 nM at α2A and α2B-adrenergic receptors respectively). Increases hippocampal noradrenalin release following systemic administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681806-46-2
Molecular Formula C17H21N3O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 331.37
Purity >98%
Molecular Weight (g/mol) 331.372
Chemical Name or Material BRL 44408 maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.