missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRL 44408 maleate, Tocris Bioscience™
Selective α2A antagonist
£169.54 - £629.69
Chemical Identifiers
CAS | 681806-46-2 |
---|---|
Molecular Formula | C17H21N3O4 |
Molecular Weight (g/mol) | 331.372 |
InChI Key | DDIQGSUEJOOQQQ-BTJKTKAUSA-N |
Synonym | brl-44408 maleate, brl 44408 maleate salt, 2-2h-1-methyl-1,3-dihydroisoindole methyl-4,5-dihydroimidazole maleate salt, brl-44408 maleate salt, brl 44408 maleate salt hplc, 2-4,5-dihydro-1h-imidazol-2-ylmethyl-1-methyl-1,3-dihydroisoindole; maleic acid, 1h-isoindole, 2-4,5-dihydro-1h-imidazol-2-yl methyl-2,3-dihydro-1-methyl-, 2z-2-butenedioate 1:1 |
PubChem CID | 10382026 |
IUPAC Name | (Z)-but-2-enedioic acid;2-(4,5-dihydro-1H-imidazol-2-ylmethyl)-1-methyl-1,3-dihydroisoindole |
SMILES | CC1C2=CC=CC=C2CN1CC3=NCCN3.C(=CC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30002138
|
Tocris Bioscience
1133/10 |
10 mg |
£169.54
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18065122
|
Tocris Bioscience
1133/50 |
50 mg |
£629.69
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective α2A-adrenoceptor antagonist (Ki = 1.7 nM and 144.5 nM at α2A and α2B-adrenergic receptors respectively). Increases hippocampal noradrenalin release following systemic administration. Also available as part of the α2-Adrenoceptor Tocriset™ .Chemical Identifiers
681806-46-2 | |
331.372 | |
brl-44408 maleate, brl 44408 maleate salt, 2-2h-1-methyl-1,3-dihydroisoindole methyl-4,5-dihydroimidazole maleate salt, brl-44408 maleate salt, brl 44408 maleate salt hplc, 2-4,5-dihydro-1h-imidazol-2-ylmethyl-1-methyl-1,3-dihydroisoindole; maleic acid, 1h-isoindole, 2-4,5-dihydro-1h-imidazol-2-yl methyl-2,3-dihydro-1-methyl-, 2z-2-butenedioate 1:1 | |
(Z)-but-2-enedioic acid;2-(4,5-dihydro-1H-imidazol-2-ylmethyl)-1-methyl-1,3-dihydroisoindole |
C17H21N3O4 | |
DDIQGSUEJOOQQQ-BTJKTKAUSA-N | |
10382026 | |
CC1C2=CC=CC=C2CN1CC3=NCCN3.C(=CC(=O)O)C(=O)O |
Specifications
681806-46-2 | |
brl-44408 maleate, brl 44408 maleate salt, 2-2h-1-methyl-1,3-dihydroisoindole methyl-4,5-dihydroimidazole maleate salt, brl-44408 maleate salt, brl 44408 maleate salt hplc, 2-4,5-dihydro-1h-imidazol-2-ylmethyl-1-methyl-1,3-dihydroisoindole; maleic acid, 1h-isoindole, 2-4,5-dihydro-1h-imidazol-2-yl methyl-2,3-dihydro-1-methyl-, 2z-2-butenedioate 1:1 | |
CC1C2=CC=CC=C2CN1CC3=NCCN3.C(=CC(=O)O)C(=O)O | |
331.372 | |
331.37 | |
BRL 44408 maleate |
C17H21N3O4 | |
DDIQGSUEJOOQQQ-BTJKTKAUSA-N | |
(Z)-but-2-enedioic acid;2-(4,5-dihydro-1H-imidazol-2-ylmethyl)-1-methyl-1,3-dihydroisoindole | |
10382026 | |
>98% |
Safety and Handling
Recommended Storage : Desiccate at -20°C