missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BRL 37344, sodium salt

Product Code. 18085362
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085362 50 mg 50mg
18064582 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085362 Supplier Tocris Bioscience™ Supplier No. 0948/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β3 agonist

Potent and selective β3 adrenoceptor agonist (Ki values are 287, 1750 and 1120 nM for β3, β1 and β2 receptors respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127299-93-8
Molecular Formula C19H21ClNNaO4

Specifications

Quantity 50 mg
Formula Weight 385.82
Purity >98%
Molecular Weight (g/mol) 385.82
Chemical Name or Material BRL 37344, sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.