missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRL-15572 dihydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
BRL-15572 dihydrochloride is a selective antagonist of h5-HT1D, displays high affinity for h5-HT1D receptors. BRL-15572 dihydrochloride could be useful pharmacological agents to characterise 5-HT1D receptor mediated responses.
Specifications
Specifications
| Chemical Name or Material | BRL-15572 dihydrochloride |
| Physical Form | Solid |
| CAS | 193611-72-2 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C25H29Cl3N2O |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 250 mg/mL (520.97 mM; Need ultrasonic) ∣H2O : 2 mg/mL (4.17 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | OC(CN1CCN(C2=CC(Cl)=CC=C2)CC1)C(C3=CC=CC=C3)C4=CC=CC=C4.[H]Cl.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction