missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brk Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98347-100ul
This item is not returnable.
View return policy
Description
Brk Polyclonal antibody specifically detects Brk in Mouse samples. It is validated for ELISA
Specifications
| Brk | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Breast tumor kinase, BRKprotein-tyrosine kinase 6, EC 2.7.10, EC 2.7.10.2, FLJ42088, protein-tyrosine kinase BRK, PTK6 protein tyrosine kinase 6, Tyrosine-protein kinase BRK | |
| Produced in rabbits immunized with purified, recombinant Mouse Brk (Uniprot#: Q64434; Met1-Val451) | |
| 100 μg | |
| Protein Kinase | |
| 5753 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction