missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRIX Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99257CL1
This item is not returnable.
View return policy
Description
BRIX Polyclonal antibody specifically detects BRIX in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| BRIX | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| brix domain containing 2, Brix domain-containing protein 2, BRIXBXDC2, BRX1, biogenesis of ribosomes, homolog (S. cerevisiae), FLJ11100, ribosome biogenesis protein BRX1 homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human BRIX. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55299 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction