missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BRIP1/FANCJ Polyclonal antibody specifically detects BRIP1/FANCJ in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
Specifications
| Antigen | BRIP1/FANCJ |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | ATP-dependent RNA helicase BRIP1, BACH1BRCA1/BRCA2-associated helicase 1, BRCA1 interacting protein C-terminal helicase 1, BRCA1-associated C-terminal helicase 1, BRCA1-binding helicase-like protein BACH1, BRCA1-interacting protein 1, BRCA1-interacting protein C-terminal helicase 1, EC 3.6.1, EC 3.6.4.13, FANCJFanconi anemia group J protein, FLJ90232, MGC126521, MGC126523, OF, Protein FACJ |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the amino acids: FNKQTKRVSWSSFNSLGQYFTGKIPKATPELGSSENSASSPPRFKTEKMESKTVLPFTDKCESSNLTVNTSFGSCPQSETIISSLKIDATLTRKNHSEHPLCSEEALDPDIELSLVSEEDKQSTSNRDFETEAEDESIYFTPEL |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?