Learn More
Invitrogen™ BRINP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588236
Description
Immunogen sequence: VQPSHQEPAG TDQHVSKEFD WLISDRGPFH HSRSYLSFVE RHRQGFTTRY KIYREFARWK VRNTAIERRD LVRHPVPLMP EFQRSIRLLG RRPTTQQFID TIIKKYGTHL LISATLGGEE ALTMYMDKSR LDRKSGNATQ SVEALHQLAS SYFVDRDGTM RRLHEIQIST GAIKVTETRT GPLGCNSYDN LDSVSSVLLQ STESKLHLQG LQIIFPQYLQ EKFVQSALSY IMCNGEGEYL CQNSQCRCQC AEEFPQCNCP ITDIQIMEYT LANMAKSWAE A; Positive Samples: U-87MG; Cellular Location: Cytoplasm.
This gene is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.
Specifications
| BRINP1 | |
| Polyclonal | |
| Unconjugated | |
| Brinp1 | |
| bA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)); BMP/retinoic acid inducible neural specific 1; BMP/retinoic acid-inducible neural-specific protein (BRINP); BMP/retinoic acid-inducible neural-specific protein 1; BMP/retinoic acid-inducible neural-specific protein-1; bone morphogenetic protein/retinoic acid inducible neural-specific 1; bone morphogenic protein/retinoic acid inducible neural specific 1; bone morphogenic protein/retinoic acid inducible neural-specific 1; Brinp; Brinp1; Dbc1; Dbccr1; deleted in bladder cancer 1; deleted in bladder cancer chromosome region candidate 1; deleted in bladder cancer protein 1; Deleted in bladder cancer protein 1 homolog; Fam5a; IB3089A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1620 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.08 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O60477 | |
| Brinp1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human BRINP1 (NP_0554332). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.