missing translation for 'onlineSavingsMsg'
Learn More

Brimonidine tartrate, Thermo Scientific™

Product Code. 17535555 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17535555 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17535555 Supplier Thermo Scientific Acros Supplier No. 468842500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Packaging Glass Bottle
CAS 70359-46-5
Certificate of Analysis Available
Physical Form Powder
Molecular Formula C15H16BrN5O6
MDL Number 07773072
Color White to Light Yellow to Light Brown
Melting Point 207°C
Solubility Soluble in water
InChI Key QZHBYNSSDLTCRG-LREBCSMRSA-N
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
Molecular Weight (g/mol) 442.23
Quantity 250 mg
Chemical Name or Material Brimonidine tartrate
Show More Show Less
compliance-icons
Product Identifier
  • Brimonidine tartrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Brimonidine tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.