missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brimonidine Tartrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Brimonidine Tartrate
Specifications
Specifications
| Chemical Name or Material | Brimonidine tartrate |
| CAS | 70359-46-5 |
| Analyte or Component Names | Brimonidine tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H10 Br N5 . C4 H6 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Brimonidine tartarate, Brimonidine D-tartrate, Brimonidine Tartrate, 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine (2R,3R)-2,3-dihydroxybutanedioate (1:1), 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), AGN 190342LF, Alphagan, Alphagan P, Brimonidine Purite, Brimonidine tartrate, Brominide tartrate |
| InChI Formula | InChI=1S/C11H10BrN5.C4H6O6/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8;5-1(3(7)8)2(6)4(9)10/h1-4H,5-6H2,(H2,15,16,17);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1 |
| SMILES | O[C@H]([C@@H](O)C(=O)O)C(=O)O.Brc1c(ccc2nccnc12)N=C3NCCN3 |
| IUPAC Name | N-(5-bromoquinoxalin-6-yl)imidazolidin-2-imine;(2R,3R)-2,3-dihydroxybutanedioic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction