missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brimonidine Tartrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Brimonidine tartrate |
| CAS | 70359-46-5 |
| Analyte or Component Names | Brimonidine tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H10 Br N5 . C4 H6 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Brimonidine tartarate, Brimonidine D-tartrate, Brimonidine Tartrate, 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine (2R,3R)-2,3-dihydroxybutanedioate (1:1), 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), AGN 190342LF, Alphagan, Alphagan P, Brimonidine Purite, Brimonidine tartrate, Brominide tartrate |
| InChI Formula | InChI=1S/C11H10BrN5.C4H6O6/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8;5-1(3(7)8)2(6)4(9)10/h1-4H,5-6H2,(H2,15,16,17);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1 |
| Solution Type | Neat |
| SMILES | O[C@H]([C@@H](O)C(=O)O)C(=O)O.Brc1c(ccc2nccnc12)N=C3NCCN3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Individuate un'opportunità di miglioramento?