missing translation for 'onlineSavingsMsg'
Learn More

Brimonidine, MedChemExpress

Product Code. 16474437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474437 50 mg 50mg
16468406 100 mg 100mg
16458568 10 mg 10mg
16421038 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474437 Supplier Medchem Express Supplier No. HYB0659
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Brimonidine (UK 14304) is a full α2-adrenergic receptor (α2-AR) agonist.

Brimonidine (UK 14304) is a full α2-adrenergic receptor (α2-AR) agonist.

Specifications

Chemical Name or Material Brimonidine
CAS 59803-98-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H10BrN5
Quantity 50 mg
Synonym UK 14304 AGN190342
Solubility Information DMSO : 50 mg/mL (171.16 mM; Need ultrasonic)
SMILES BrC1=C2N=CC=NC2=CC=C1NC3=NCCN3
Molecular Weight (g/mol) 292.13
Formula Weight 292.13
Percent Purity 99.09%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.