missing translation for 'onlineSavingsMsg'
Learn More

Brimonidine-2,3-dione, TRC

Product Code. 30137575
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137575 100 mg 100mg
30007747 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137575 Supplier Toronto Research Chemicals Supplier No. TRCB677525100MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182627-95-8
Molecular Formula C11 H10 Br N5 O2
Molecular Weight (g/mol) 324.13
Synonym 2,3-Quinoxalinedione, 5-bromo-6-[(4,5-dihydro-1H-imidazol-2-yl)amino]-1,4-dihydro-, 5-Bromo-6-[(4,5-dihydro-1H-imidazol-2-yl)amino]-1,4-dihydro-2,3-quinoxalinedione, 5-Bromo-6-[(4,5-dihydro-1H-imidazol-2-yl)amino]-1,4-dihydroquinoxaline-2,3-dione
IUPAC Name 5-bromo-6-(4,5-dihydro-1H-imidazol-2-ylamino)-1,4-dihydroquinoxaline-2,3-dione
SMILES Brc1c(NC2=NCCN2)ccc3NC(=O)C(=O)Nc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H10BrN5O2/c12-7-5(16-11-13-3-4-14-11)1-2-6-8(7)17-10(19)9(18)15-6/h1-2H,3-4H2,(H,15,18)(H,17,19)(H2,13,14,16)
Formula Weight 323.0018
Chemical Name or Material Brimonidine-2,3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.