missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Cresyl blue (Technical Grade), TRC

Product Code. 30342650
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342650 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342650 Supplier Toronto Research Chemicals Supplier No. TRCB67741325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brilliant Cresyl blue (Technical Grade)
CAS 81029-05-2
Molecular Formula 2[C17 H21 N4 O]. Cl4 Zn
Quantity 25 g
Synonym 1,3-Diamino-7-(diethylamino)-4-methyl-phenoxazin-5-ium (T-4)-Tetrachlorozincate(2-)
InChI Formula InChI=1S/2C17H21N4O.4ClH.Zn/c2*1-4-21(5-2)11-6-7-14-15(8-11)22-17-10(3)12(18)9-13(19)16(17)20-14;;;;;/h2*6-9H,4-5,18-19H2,1-3H3;4*1H;/q2*+1;;;;;+2/p-4
SMILES [Cl-][Zn+2]([Cl-])([Cl-])[Cl-].CCN(CC)c1ccc2nc3c(N)cc(N)c(C)c3[o+]c2c1.CCN(CC)c4ccc5nc6c(N)cc(N)c(C)c6[o+]c5c4
IUPAC Name mono(1,3-diamino-7-(diethylamino)-4-methylphenoxazin-5-ium) mono(tetrachlorozincate(II))
Molecular Weight (g/mol) 801.97068
Formula Weight 798.1476
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.