missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant cresyl Blue, pure, high purity biological stain

Product Code. 10480881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10480881 25 g Glass bottle 25g
10338410 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10480881 Supplier Thermo Scientific Chemicals Supplier No. 229590250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81029-05-2 | 0·5 ZnCl4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81029-05-2
Molecular Formula 0·5 ZnCl4
MDL Number MFCD00148902

Specifications

Absorbance 1500 (625 to 630nm, 50% min. ethanol) (A (1% min./1cm))
Infrared Spectrum Authentic
Formula Weight 400.97
Loss on Drying 8 % max. (1 g, 110°C)
Grade Pure
Packaging Glass bottle
Specific Extinction 1500 to 1700 (622nm, 50% ethanol)
Color White
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Brilliant cresyl blue
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.