missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Blue G (Technical Grade), TRC

Product Code. 30341209
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30341209

Brand: Toronto Research Chemicals TRCB67740310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brilliant Blue G
CAS 6104-58-1
Molecular Formula C47 H49 N3 O7 S2 . Na
Quantity 10 g
Synonym Brilliant Indocyanine G, C.I. Acid Blue 90, C.I. Acid Blue 90, Monosodium Salt, Acid Blue 90, Acid Blue G 4061, Acid Blue PG, Acid Bright Blue G, Acid Brilliant Blue G, Acid Brilliant Cyanine G, Acidine Sky Blue G, Amacid Brilliant Cyanine G, Anadurm Cyanine A-G, BBG, Benzyl Cyanine G, Biosafe Coomassie Stain, Brilliant Acid Blue G, Brilliant Acid Blue GI, Brilliant Acid Blue J, Brilliant Acid Cyanine G, Brilliant Blue G 250, Brilliant Blue J, Brilliant Indocyanine GA-CF, Bucacid Brilliant Indocyanine G, C.I. 42655, CBB-G 250, Colocid Brilliant Blue EG, Coomassie Blue G, Coomassie Blue G 250, Coomassie Brilliant Blue G, Coomassie Brilliant Blue G 250, Coomassie G 250, Cyanine G, Daiwa Acid Blue 300, Derma Cyanine G, Derma Cyanine GN 360, Dycosweak Acid Brilliant Blue G, Eriosin Brilliant
InChI Formula InChI=1S/C47H49N3O7S2.Na/c1-6-49(31-35-11-9-13-43(29-35)58(51,52)53)40-21-25-45(33(4)27-40)47(37-15-17-38(18-16-37)48-39-19-23-42(24-20-39)57-8-3)46-26-22-41(28-34(46)5)50(7-2)32-36-12-10-14-44(30-36)59(54,55)56;/h9-30H,6-8,31-32H2,1-5H3,(H2,51,52,53,54,55,56);/q;+1
SMILES [Na+].CCOc1ccc(Nc2ccc(cc2)\C(=C\3/C=C\C(=[N+](\CC)/Cc4cccc(c4)S(=O)(=O)[O-])\C=C3C)\c5ccc(cc5C)N(CC)Cc6cccc(c6)S(=O)(=O)O)cc1
IUPAC Name sodium;3-[[(E)-[(4Z)-4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfophenyl)methyl]amino]-2-methyl-phenyl]methylene]-3-methyl-cyclohexa-2,5-dien-1-ylidene]-ethyl-ammonio]methyl]benzenesulfonate
Molecular Weight (g/mol) 855.03
Formula Weight 854.2904
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.