missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Blue G, pure

Product Code. 15710829 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15710829 100 g Glass bottle 100g
10266390 5 g Glass bottle 5g
10358090 25 g Glass bottle 25g
10398933 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15710829 Supplier Thermo Scientific Chemicals Supplier No. 191481000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6104-58-1 | C47H48N3NaO7S2 | 854.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
Molecular Weight (g/mol) 854.025
MDL Number MFCD00078482
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Synonym Acid Blue 90, Brilliant Blue G 250, C.I. 42655
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]

Specifications

Absorbance 425 (at lambda min.)
Melting Point >100.0°C
Absorption Ratio 0.95 to 1.15
Dye Content 60% min.
Infrared Spectrum Authentic
Lambda Max 608 to 618nm (in 50:50 IMS:H2O)
Loss on Drying 10% max. (105°C)
Packaging Glass bottle
Beilstein 14, II, 450
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: soluble in methanol, ethanol
Quantity 100 g
Physical Form Crystalline Powder
Formula Weight 854.02
Grade Pure
Chemical Name or Material Brilliant blue G
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.