missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Black BN, MP Biomedicals™

Product Code. 11477972 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477972 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477972 Supplier MP Biomedicals Supplier No. 0215263305
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2519-30-4
Molecular Formula C28H17N5Na4O14S4
Molecular Weight (g/mol) 867.664
InChI Key AGKKBBSOKGLVTM-YENKNCRGSA-J
Synonym CI 28440, Food Black 1
PubChem CID 6321379
IUPAC Name tetrasodium;(6E)-4-acetamido-5-oxo-6-[[7-sulfonato-4-[(4-sulfonatophenyl)diazenyl]naphthalen-1-yl]hydrazinylidene]naphthalene-1,7-disulfonate
SMILES CC(=O)NC1=C2C(=C(C=C1)S(=O)(=O)[O-])C=C(C(=NNC3=C4C=C(C=CC4=C(C=C3)N=NC5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-])C2=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Formula Weight 867.7
Color Black
Quantity 5 g
Physical Form Powder
Chemical Name or Material Brilliant Black BN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.