missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance UTI Medium

Product Code. 15839502
Change view
Click to view available options
Unit Size:
400g
Each
Quantity:
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Quantity
15839502 400g -
15823217 Each 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15839502 Supplier Thermo Scientific™ Supplier No. CM0949C
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Brilliance™ UTI Agar devices are differential media for the isolation and presumptive differentiation of the common microorganisms causing urinary tract infections (UTIs) from clinical samples, including urine samples.

Urinary tract infection (UTI) occurs due to the presence and growth of microorganisms in the urinary tract, giving rise to a range of clinical presentations. Prominent human pathogens are often associated with UTIs, with the most common microorganism involved in urinary tract infections being E. coli, accounting for around 77% of isolates. Other microorganisms associated with acute, uncomplicated UTIs include Proteus mirabilis, Staphylococcus saprophyticus, other coagulase negative staphylococci, streptococci, and Candida species. Other microorganisms are associated with complicated UTIs, including Klebsiella, Enterobacter and Proteus species, Enterococcus species and Pseudomonas aeruginosa. Although S. aureus rarely causes infection, it is associated with renal abnormality, secondary infections to bacteraemia, surgery, and catheterisation. Other coagulase negative Staphylococci have been associated with complicated infections in individuals with abnormalities of the urinary tract or underlying disease.

Differentiation of the common microorganisms causing urinary tract infections (UTIs) is achieved through the inclusion of two chromogens that are targeted by specific enzymes: β-galactosidase and β- glucosidase. The action of these enzymes on the chromogens causes release of the coloured component inside the bacterial cell, resulting in coloured colonies. The colour produced depends on which enzymes the organisms produce. β-galactosidase is produced by E. coli, which grow as pink colonies. β- glucosidase is produced by enterococci which form blue or turquoise colonies. Cleavage of both chromogens by the KESC (Klebsiella, Enterobacter, Serratia, Citrobacter) group of coliforms gives dark blue or purple colonies. Tryptophan acts as an indicator of tryptophan deaminase activity (TDA), resulting in colonies of Proteus, Morganella and Providencia spp. with brown halos. Peptone forms the nutritional source within the medium.

Brilliance™ UTI Agar devices are used in a diagnostic workflow to aid clinicians in determining potential treatment options for patients suspected of having UTIs. The devices are for professional use only, are not automated and nor are they companion diagnostics.
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated Powder
Manufacturing Origin United Kingdom
Sterility Non-sterile
Target Organism Class Bacterial Pathogens
Test Type Microbial Differentiation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.