Learn More
Bright Brushing Gold, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 012943.04
Code : NEW
Additional Details : Weight : 0.00200kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Red to Brown | |
156°C | |
UN1993 | |
33°C (91°F) | |
Liquid |
Bright Brushing Gold | |
MFCD00003436 | |
2 g | |
Pleasant |
RUO – Research Use Only