missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Briakinumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Briakinumab |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30282631
![]() |
Invitrogen™
MA559519 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30284191
|
Invitrogen™
MA559520 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Briakinumab is a human anti-IL-12 monoclonal antibody being developed for the treatment of a number of T-cell driven autoimmune diseases. It targets and neutralize interleukin-12 and interleukin-23, two proteins associated with inflammation, such as pro-inflammatory interleukins or tumor necrosis factor- alpha. Briakinumab represents a novel approach to treating psoriasis, Multiple Sclerosis, Crohn's Disease and other autoimmune and inflammatory disorders. As of 2011 pharmaceutical development for psoriasis has been discontinued in the U. S. and Europe. Briakinumab targets and neutralizes interleukin-12 and interleukin-23.Specifications
| Briakinumab | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| ABT-874 | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title