missing translation for 'onlineSavingsMsg'
Learn More

Brg1 Antibody, FITC, Novus Biologicals™

Product Code. 18401352 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401352 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401352 Supplier Novus Biologicals Supplier No. NBP222234F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Brg1 Polyclonal antibody specifically detects Brg1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brg1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATP-dependent helicase SMARCA4, BAF190, BAF190A, brahma protein-like 1, BRG1-associated factor 190A, BRG1SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 4, BRM/SWI2-related gene 1, EC 3.6.1, EC 3.6.4.-, FLJ39786, global transcription activator homologous sequence, homeotic gene regulator, hSNF2b, Mitotic growth and transcription activator, nuclear protein GRB1, Protein brahma homolog 1, Protein BRG-1, RTPS2, SNF2, SNF2B, SNF2-BETA, SNF2L4, SNF2LB, SNF2-like 4, subfamily a, member 4, sucrose nonfermenting-like 4, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI2, transcription activator BRG1
Host Species Goat
Immunogen The immunogen this antibody was made to, maps to a region between residue 50 and 100 of human BRM/SWI2-related Gene 1 using the numbering given in entry NP_003063.2 (GeneID 6597).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6597
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.