missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRF2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92838-0.1ml
This item is not returnable.
View return policy
Description
BRF2 Polyclonal antibody specifically detects BRF2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BRF2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| B-related factor 2, BRF-2, BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like, BRFUhTFIIIB50, FLJ11052, hBRFU, TFIIIB50RNA polymerase III transcription initiation factor BRFU, transcription factor IIB- related factor, TFIIIB50, transcription factor IIIB 50 kDa subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 180-419 of human BRF2 (NP_060780.2). FKLFQASPSVPAKYVEDKEKMLSRTMQLVELANETWLVTGRHPLPVITAATFLAWQSLQPADRLSCSLARFCKLANVDLPYPASSRLQELLAVLLRMAEQLAWLRVLRLDKRSVVKHIGDLLQHRQSLVRSAFRDGTAEVETREKEPPGWGQGQGEGEVGNNSLGLPQGKRPASPALLLPPCMLKSPKRICPVPPVSTVTGDENISDSEIEQYLRTPQEVRDFQRAQAARQAATSVPNPP | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55290 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction