missing translation for 'onlineSavingsMsg'
Learn More

BRF1 Antibody, Novus Biologicals™

Product Code. 18373988 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373988 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373988 Supplier Novus Biologicals Supplier No. H00002972B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

BRF1 Polyclonal antibody specifically detects BRF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_663718.1
Gene Alias B-related factor 1, BRF-1, BRF1 homolog, subunit of RNA polymerase III transcription initiation factorIIIB (S. cerevisiae), BRFFLJ42674, general transcription factor IIIB, 90kD subunit, GTF3BTAFIII90, hBRFMGC105048, hTFIIIB90, TAF3B2, TAF3CB - related factor 1, TATA box binding protein (TBP)-associated factor 3C, TATA box binding protein (TBP)-associated factor, RNA polymerase III, GTF3Bsubunit 2, TATA box binding protein (TBP)-associated factor, RNA polymerase III, subunit 2, TATA box-binding protein-associated factor, RNA polymerase III, subunit 2, TBP - associated factor, RNA polymerase III, 90kD, TF3B90, TFIIIB90FLJ43034, transcription factor IIIB 90 kDa subunit
Host Species Mouse
Immunogen BRF1 (NP_663718.1, 1 a.a. - 473 a.a.) full-length human protein. MTALRLLQRMKRDWMHTGRRPSGLCGAALLVAARMHDFRRTVKEVISVVKVCESTLRKRLTEFEDTPTSQLTIDEFMKIDLEEECDPPSYTAGQRKLRMKQLEQVLSKKLEEVEGEISSYQDAIEIELENSRPKAKGGLASLAKDGSTEDTASSLCGEEDTEDEELEAAASHLNKDLYRELLGGAPGSSEAAGSPEWGGRPPALGSLLDPLPTAASLGISDSIRECISSQSSDPKDASGDGELDLSGIDDLEIDRYILNESEARVKAELWMRENAEYLREQREKEARIAKEKELGIYKEHKPKKSCKRREPIQASTAREAIEKMLEQKKISSKINYSVLRGLSSAGGGSPHREDAQPEHSASARKLSRRRTPASRSGADPVTSVGKRLRPLVSTQPAKKVATGEALLPSSPTLGAEPARPQAVLVESGPVSYHADEEADEEEPDEEDGEPCVSALQMMGSNDYGCDGDEDDGY
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2972
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.