missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Brevican Polyclonal Antibody

Rabbit Polyclonal Antibody
£178.00 - £430.00
Specifications
| Antigen | Brevican |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17790394
![]() |
Invitrogen™
PA5118986 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17763584
|
Invitrogen™
PA5122188 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Brevican Polyclonal Antibody for Western Blot, IHC (P)
Brevican is the most abundant chondroitin sulfate proteoglycan in the extracellular matrix of the adult brain. It is a member of the lectican family of aggregating extracellular matrix proteoglycans that bear chondroitin sulfate (CS) chains. It is highly expressed in the central nervous system and is thought to stabilize synapses and inhibit neural plasticity. Brevican is secreted from astrocytes and neurons as a 145 kD core protein that bears up to three, covalently-linked, CS chains. It is also is secreted as a 145 kD core protein without CS chains. When cleaved by extracellular glutamyl endopeptidases, the ADAMTSs, a 55 kD N-terminal fragment is formed that contains the unique C terminal epitope EAMESE.Specifications
| Brevican | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| Bcan | |
| ALPBRE; Bcan; Behab; Brain-enriched hyaluronan-binding protein; brevican; brevican (brain specific proteoglycan in the aggrecan family); brevican core protein; Chondroitin sulfate proteoglycan 7; chondroitin sulfate proteoglycan BEHAB; CSPG7; UNQ2525/PRO6018 | |
| Bcan | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P55068, Q61361 | |
| 12032, 25393 | |
| Recombinant protein Brevican. The antigen corresponds to amino acid range 23-353 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title