missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Brevican Monoclonal Antibody (450613)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524170
This item is not returnable.
View return policy
Description
This antibody also detects a truncated version of recombinant human Brevican (aa 23-649). Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Brevican is the most abundant chondroitin sulfate proteoglycan in the extracellular matrix of the adult brain. It is a member of the lectican family of aggregating extracellular matrix proteoglycans that bear chondroitin sulfate (CS) chains. It is highly expressed in the central nervous system and is thought to stabilize synapses and inhibit neural plasticity. Brevican is secreted from astrocytes and neurons as a 145 kD core protein that bears up to three, covalently-linked, CS chains. It is also is secreted as a 145 kD core protein without CS chains. When cleaved by extracellular glutamyl endopeptidases, the ADAMTSs, a 55 kD N-terminal fragment is formed that contains the unique C terminal epitope EAMESE.
Specifications
| Brevican | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q96GW7 | |
| Bcan | |
| Mouse myeloma cell line NS0-derived recombinant human Brevican isoform 1 Asp23-Pro911. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Neutralization, Western Blot | |
| 450613 | |
| Unconjugated | |
| Bcan | |
| ALPBRE; Bcan; Behab; Brain-enriched hyaluronan-binding protein; brevican; brevican (brain specific proteoglycan in the aggrecan family); brevican core protein; Chondroitin sulfate proteoglycan 7; chondroitin sulfate proteoglycan BEHAB; CSPG7; UNQ2525/PRO6018 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 63827 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction