missing translation for 'onlineSavingsMsg'
Learn More

Bretylium Tosylate, TRC

Product Code. 30345439
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30345439

Brand: Toronto Research Chemicals TRCB67737010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bretylium Tosylate
CAS 61-75-6
Molecular Formula C11 H17 Br N . C7 H7 O3 S
Quantity 10 mg
Synonym Benzenemethanaminium, 2-bromo-N-ethyl-N,N-dimethyl-, 4-methylbenzenesulfonate (1:1), (o-Bromobenzyl)ethyldimethylammonium p-toluenesulfonate (6CI,7CI), Ammonium, (o-bromobenzyl)ethyldimethyl-, p-toluenesulfonate (8CI), Benzenemethanaminium, 2-bromo-N-ethyl-N,N-dimethyl-, salt with 4-methylbenzenesulfonic acid (1:1) (9CI), ASL 603, Bretylan, Bretylate, Bretylium p-toluenesulfonate, Bretylium tosilate, Bretylium tosylate, Bretylol, Darenthin, N-Ethyl-N-(o-bromobenzyl)-N,N-dimethylammonium tosylate, NSC 62164, Ornid
InChI Formula InChI=1S/C11H17BrN.C7H8O3S/c1-4-13(2,3)9-10-7-5-6-8-11(10)12;1-6-2-4-7(5-3-6)11(8,9)10/h5-8H,4,9H2,1-3H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1
SMILES CC[N+](C)(C)Cc1ccccc1Br.Cc2ccc(cc2)S(=O)(=O)[O-]
IUPAC Name (2-bromophenyl)methyl-ethyl-dimethylazanium;4-methylbenzenesulfonate
Molecular Weight (g/mol) 414.36
Formula Weight 413.066
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.