missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bretylium Tosylate, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Bretylium Tosylate |
| CAS | 61-75-6 |
| Molecular Formula | C11 H17 Br N . C7 H7 O3 S |
| Quantity | 100 mg |
| Synonym | Benzenemethanaminium, 2-bromo-N-ethyl-N,N-dimethyl-, 4-methylbenzenesulfonate (1:1), (o-Bromobenzyl)ethyldimethylammonium p-toluenesulfonate (6CI,7CI), Ammonium, (o-bromobenzyl)ethyldimethyl-, p-toluenesulfonate (8CI), Benzenemethanaminium, 2-bromo-N-ethyl-N,N-dimethyl-, salt with 4-methylbenzenesulfonic acid (1:1) (9CI), ASL 603, Bretylan, Bretylate, Bretylium p-toluenesulfonate, Bretylium tosilate, Bretylium tosylate, Bretylol, Darenthin, N-Ethyl-N-(o-bromobenzyl)-N,N-dimethylammonium tosylate, NSC 62164, Ornid |
| InChI Formula | InChI=1S/C11H17BrN.C7H8O3S/c1-4-13(2,3)9-10-7-5-6-8-11(10)12;1-6-2-4-7(5-3-6)11(8,9)10/h5-8H,4,9H2,1-3H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1 |
| SMILES | CC[N+](C)(C)Cc1ccccc1Br.Cc2ccc(cc2)S(=O)(=O)[O-] |
| IUPAC Name | (2-bromophenyl)methyl-ethyl-dimethylazanium;4-methylbenzenesulfonate |
| Molecular Weight (g/mol) | 414.36 |
| Formula Weight | 413.066 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction