missing translation for 'onlineSavingsMsg'
Learn More

Bretylium tosylate, MedChemExpress

Product Code. 16495448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495448 10 mg 10mg
16471627 50 mg 50mg
16451056 200 mg 200mg
16438167 5 mg 5mg
16436046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495448 Supplier Medchem Express Supplier No. HY12961A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bretylium (tosylate) is an inhibitor of the presynaptic release of vasoconstrictor neurotransmitters.

Bretylium (tosylate) is an inhibitor of the presynaptic release of vasoconstrictor neurotransmitters.

Specifications

Chemical Name or Material Bretylium tosylate
CAS 61-75-6
Color White
Purity Grade Notes Research
Molecular Formula C18H24BrNO3S
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (72.40 mM)
SMILES C[N+](C)(CC)CC1=CC=CC=C1Br.O=S(C2=CC=C(C)C=C2)([O-])=O
Molecular Weight (g/mol) 414.36
Formula Weight 414.36
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.