missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Bretazenil

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Bretazenil is a partial agonist at the GABAA benzodiazepine site (EC50 = 10 nM at α1β1γ2 receptors). Displays anticonvulsive activity in vivo.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | Bretazenil |
| Molecular Weight (g/mol) | 418.28 |
| Inhibitors | Bretazenil |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction