missing translation for 'onlineSavingsMsg'
Learn More

Brepocitinib P-Tosylate, MedChemExpress

Product Code. 16485516
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485516 100 mg 100mg
16477397 5 mg 5mg
16424688 10 mg 10mg
16420977 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485516 Supplier Medchem Express Supplier No. HY112708A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Brepocitinib (PF-06700841) P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. Brepocitinib P-Tosylate also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively.

Brepocitinib (PF-06700841) P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. Brepocitinib P-Tosylate also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively.

Specifications

Chemical Name or Material Brepocitinib P-Tosylate
CAS 2140301-96-6
Color White
Purity Grade Notes Research
Molecular Formula C25H29F2N7O4S
Quantity 100 mg
Synonym PF-06700841 P-Tosylate
Solubility Information DMSO : 62.5 mg/mL (111.29 mM; Need ultrasonic)
SMILES O=C([C@H]1C(F)(F)C1)N2C3CN(C4=NC(NC5=CN(C)N=C5)=NC=C4)CC2CC3.O=S(C6=CC=C(C)C=C6)(O)=O
Molecular Weight (g/mol) 561.6
Formula Weight 561.6
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.