missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bremelanotide Acetate

Product Code. 16457578
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16457578 Supplier MedChemExpress Supplier No. HY18678A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bremelanotide Acetate (PT-141 Acetate), a synthetic peptide analogue of α-MSH, is an agonist at melanocortin receptors including the MC3R and MC4R for the treatment of sexual dysfunction.

Specifications

Purity 98.18%
CAS 1607799-13-2
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 1085.22 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 36 mg/mL (33.17 mM)
Synonym PT-141 Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.