missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brefeldin A, natural

Catalog Number 10350834
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
20350-15-6
C16H24O4
280.36
MFCD00083258,MFCD12913297
KQNZDYYTLMIZCT-PNFJWZTBSA-N
brefeldin a, cyanein, decumbin, bfa
133640201
(1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one
C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
20350-15-6
C16H24O4
280.36
MFCD00083258,MFCD12913297
KQNZDYYTLMIZCT-PNFJWZTBSA-N
brefeldin a, cyanein, decumbin, bfa
133640201
(1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one
C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 201.0°C to 205.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 mg
Merck Index 15, 1374
Specific Rotation + 93.00
Solubility Information Solubility in water: .. Other solubilities: 10 mg/mL methanol
Formula Weight 280.35
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material Brefeldin A
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'