missing translation for 'onlineSavingsMsg'
Learn More

Brefeldin A, 98%

Product Code. 15484739
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484739 50 mg 50mg
15464739 10 mg 10mg
15474739 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15484739 Supplier Thermo Scientific Alfa Aesar Supplier No. J62340.MF
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20350-15-6 | C16H24O4 | 280.36 g/mol

Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

Solubility
Soluble in dimethylsulfoxide, dichloromethane and ethanol. Slightly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20350-15-6
Molecular Formula C16H24O4
Molecular Weight (g/mol) 280.36
MDL Number MFCD00083258,MFCD12913297
InChI Key KQNZDYYTLMIZCT-PNFJWZTBSA-N
Synonym brefeldin a, cyanein, decumbin, bfa
PubChem CID 133640201
SMILES C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1

Specifications

Melting Point 201°C to 205°C
Color White
Quantity 50 mg
UN Number UN2811
Beilstein 25191
Merck Index 14,1369
Solubility Information Soluble in dimethylsulfoxide,dichloromethane and ethanol. Slightly soluble in water.
IUPAC Name (1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one
Formula Weight 280.36
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Brefeldin A
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.