Learn More
Greiner Bio-One BREATHseal™ Sealer
Product Code. 11920667
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
11920667 Sterile Pack of 50
11910667 Non-sterile Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11920667 Supplier Greiner Bio-One Supplier No. 676051
Add to basket
Add to basket
For the culture of prokaryotic and eukaryotic cells
  • Unique gas-permeable sealer with a wide distribution of pores for excellent gas exchange
  • Available sterile and non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Color Translucent
Material Rayon-Fibers Foil, Poly-coated Kraft Paper with Silicone
Sterility Sterile
Product Type BREATHseal Sealer
Adhesive Acrylic
Thickness 0.14 mm
Pierceable Yes
Plate Compatibility Cell Culture Microplate, HTS Microplate