missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Breast cancer suppressor candidate 1 Antibody (OTI3D6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71158AF532
This item is not returnable.
View return policy
Description
Breast cancer suppressor candidate 1 Monoclonal specifically detects Breast cancer suppressor candidate 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Breast cancer suppressor candidate 1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| VWA5A | |
| Full length recombinant protein of human Breast cancer suppressor candidate 1 (NP_055437) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3D6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A | |
| Mouse | |
| 0.1 mL | |
| 4013 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction