missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Breast cancer suppressor candidate 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | Breast cancer suppressor candidate 1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18486841
|
Novus Biologicals
NBP1-89998 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18470252
|
Novus Biologicals
NBP1-89998-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Breast cancer suppressor candidate 1 Polyclonal antibody specifically detects Breast cancer suppressor candidate 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Breast cancer suppressor candidate 1 | |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SLSWHLPPGLSAKMLSPEQTVIFRGQRLISYAQLTGRMPAAETTGEVCLKYTLQGKTFEDKVTFPLQPKPDVNLTIHRLAAKSLLQTKDMGLRETPASDK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 4013 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title