missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BRE Polyclonal antibody specifically detects BRE in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | BRE |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | brain and reproductive organ-expressed (TNFRSF1A modulator), BRCA1/BRCA2-containing complex subunit 45, subunit 4 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human BRCC45. The immunogen is located within the last 50 amino acids of BRCC45. Amino Acid Squence: YSQAQKNYPYSPRWDGNE |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?