Learn More
Invitrogen™ BrdU Monoclonal Antibody (MoBU-1), Alexa Fluor™ 647
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ B35133
Product Code. 10335988
  • £452.00 / 350µL
For Research Use Only. All usage must comply with product instructions.
Description
Description
Cell proliferation can be measured with BrdU following its incorporation into newly synthesized DNA and its subsequent detection with an anti-BrdU antibody. Although this method is rapidly being replaced with the recently developed click chemistry-based Click-iTTM EdU assay, an advantage of the antibody clone MoBU-1 is that it does not cross-react with the thymidine analog EdU (5-ethynyl-2'-deoxyuridine), thus making it valuable for use in dual-pulse applications. Traditionally, the dual-pulse method, which is used for studying biological events such as neurogenesis and cell differentiation or evaluating anti-cancer drug efficacy, employs BrdU immunocytochemistry with iododeoxyuridine (IdU) or chlorodeoxyuridine (CldU), using multiple antibodies for detection. However, using sequential pulses of the thymidine analogs EdU and BrdU greatly simplifies dual-pulse labeling, and allows a straightforward and reliable method of distinguishing BrdU- and EdU-labeled cells. In addition to no cross-reactivity with EdU, the MoBU-1 clone has a high specificity for BrdU. This clone can also readily detect bromouridine (BrU) incorporated into RNA or BrdUTP incorporated into DNA strand breaks via the TUNEL assay. Samples labeled with the Alexa FluorTM conjugates of MoBU-1 can be directly imaged by fluorescence microscopy or high-content imaging. The approximate fluorescence excitation/emission maxima for Alexa FluorTM647 conjugate are 650/665 or 670 in nm. Detection with BrdU antibodies re...
Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BrdU
Monoclonal
0.2 mg/mL
PBS with 5mM sodium azide; pH 7.2
Mouse
Affinity chromatography
RUO
Chemical
Antibody
IgG
Immunohistochemistry
MoBU-1
Alexa Fluor 647
5-Bromo-2-deoxyuridine; Bromodeoxyuridine
BrdU.
350 μL
Primary
4°C
Liquid
Chemical