Learn More
Invitrogen™ BrdU Monoclonal Antibody (BU-1)

Description
MA3-071 detects 5-Bromo-2-deoxyuridine (BrdU) in many samples. MA3-071 has been successfully used in immunofluorescence, immunohistochemistry, and flow cytometry applications. The MA3-071 immunogen is 5-iodouridine covalently coupled to ovalbumin. This antibody was orginally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.
Specifications
Specifications
| Antigen | BrdU |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | BU-1 |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 0.15M NaCl, 30% glycerol and 0.01% sodium azide |
| Gene Alias | 5-Bromo-2-deoxyuridine; Bromodeoxyuridine |
| Host Species | Mouse |
| Immunogen | 5-iodouridine covalently coupled to ovalbumin. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.