missing translation for 'onlineSavingsMsg'
Learn More

BRDG1/STAP-1 Antibody, Novus Biologicals™

Product Code. 18410700 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410700 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410700 Supplier Novus Biologicals Supplier No. H00026228D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BRDG1/STAP-1 Polyclonal antibody specifically detects BRDG1/STAP-1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRDG1/STAP-1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_036240.1
Gene Alias BCR downstream-signaling protein 1, BRDG1STAP-1BCR downstream signaling 1, Docking protein BRDG1, signal transducing adaptor family member 1, signal-transducing adaptor protein 1, signal-transducing adaptor protein-1, Stem cell adaptor protein 1
Host Species Rabbit
Immunogen STAP1 (NP_036240.1, 1 a.a. - 295 a.a.) full-length human protein. MMAKKPPKPAPRRIFQERLKITALPLYFEGFLLIKRSGYREYEHYWTELRGTTLFFYTDKKSIIYVDKLDIVDLTCLTEQNSTEKNCAKFTLVLPKEEVQLKTENTESGEEWRGFILTVTELSVPQNVSLLPGQVIKLHEVLEREKKRRIETEQSTSVEKEKEPTEDYVDVLNPMPACFYTVSRKEATEMLQKNPSLGNMILRPGSDSRNYSITIRQEIDIPRIKHYKVMSVGQNYTIELEKPVTLPNLFSVIDYFVKETRGNLRPFICSTDENTGQEPSMEGRSEKLKKNPHIA
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26228
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.