missing translation for 'onlineSavingsMsg'
Learn More

BRD9539, MedChemExpress

Product Code. 16479267
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479267 5 mg 5mg
16443486 25 mg 25mg
16442797 50 mg 50mg
16426718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16479267 Supplier Medchem Express Supplier No. HY15647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1.

BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1.

Specifications

Chemical Name or Material BRD9539
CAS 1374601-41-8
Color White
Purity Grade Notes Research
Molecular Formula C24H21N3O3
Quantity 5 mg
Solubility Information DMSO : 16.67 mg/mL (41.73 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2C(N=C(NC(C3=CC=CC=C3)=O)N2CCCC4=CC=CC=C4)=C1)O
Molecular Weight (g/mol) 399.44
Formula Weight 399.44
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.