missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD9 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | BRD9 |
|---|---|
| Concentration | 587 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16883275
|
Proteintech
24785-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16873275
|
Proteintech
24785-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BRD9 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human BRD9. This antibody recognizes human antigen. The BRD9 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BRD9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
| 587 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 65980 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title