missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD8, Mouse anti-Human, Clone: 3G8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010902-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: RSGDQNWVSVSRAIKPFAEPGRPPDWFSQKHCASQYSELLETTETPKRKRGEKGEVVETVEDVIVRKLTAERVEELKKVIKETQERYRRLKRDAELSpecifications
| BRD8 | |
| Monoclonal | |
| Unconjugated | |
| bromodomain containing 8 | |
| SMAP/SMAP2/p120 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence | |
| 3G8 | |
| In 1x PBS, pH 7.4 | |
| NM_139199 | |
| BRD8 | |
| BRD8 (NP_631938, 33 a.a. ∼ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10902 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction