missing translation for 'onlineSavingsMsg'
Learn More

BRD8 Antibody (3G8), Novus Biologicals™

Product Code. 18372019 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372019 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372019 Supplier Novus Biologicals Supplier No. H00010902M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BRD8 Monoclonal antibody specifically detects BRD8 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD8
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3G8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_631938
Gene Alias bromodomain containing 8, bromodomain-containing protein 8, Skeletal muscle abundant protein, SMAP2, SMAPp120Skeletal muscle abundant protein 2, Thyroid hormone receptor coactivating protein of 120 kDa, trCP120
Host Species Mouse
Immunogen BRD8 (NP_631938, 33 a.a. ~ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RSGDQNWVSVSRAIKPFAEPGRPPDWFSQKHCASQYSELLETTETPKRKRGEKGEVVETVEDVIVRKLTAERVEELKKVIKETQERYRRLKRDAEL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 10902
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.