missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD7552, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
BRD7552, a potent PDX1 transcription factor inducer, upregulates PDX1 expression in both primary human islets and ductal cells, and induces epigenetic changes in the PDX1 promoter consistent with transcriptional activation. BRD7552 increases insulin expression. PDX1 is a key transcription factor involved in pancreas development and β cell function.
Specifications
Specifications
| Chemical Name or Material | BRD7552 |
| CAS | 1137359-47-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C33H33N3O15 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 38 mg/mL (53.40 mM) |
| SMILES | OC[C@@H](O1)[C@@H](OC(NC2=CC=C(C(OCC)=O)C=C2)=O)[C@H](OC(NC3=CC=C(OCO4)C4=C3)=O)[C@@H](OC(NC5=CC=C(OCO6)C6=C5)=O)[C@H]1OC |
| Molecular Weight (g/mol) | 711.63 |
| Formula Weight | 711.63 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction