missing translation for 'onlineSavingsMsg'
Learn More

BRD7116, MedChemExpress

Product Code. 16487598
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487598 10 mg 10mg
16473617 50 mg 50mg
16420078 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487598 Supplier Medchem Express Supplier No. HY18714
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BRD7116 competitively binds to bacterial DNA gyrase, exhibits an EC50 of 200 nM for LSCe cells, with cell-non-autonomous anti-leukemia activity.

BRD7116 competitively binds to bacterial DNA gyrase, exhibits an EC50 of 200 nM for LSCe cells, with cell-non-autonomous anti-leukemia activity.

Specifications

Chemical Name or Material BRD7116
CAS 329059-55-4
Color White
Purity Grade Notes Research
Molecular Formula C28H36N2O4S
Quantity 10 mg
Solubility Information DMSO : ≥ 48 mg/mL (96.65 mM)
SMILES O=S(C1=CC=C(NC(C2C(C)(C)C2(C)C)=O)C=C1)(C3=CC=C(NC(C4C(C)(C)C4(C)C)=O)C=C3)=O
Molecular Weight (g/mol) 496.66
Formula Weight 496.66
Percent Purity 99.73%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.