missing translation for 'onlineSavingsMsg'
Learn More

BRD7-IN-1, MedChemExpress

Product Code. 16450847
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450847

Brand: Medchem Express HY111905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BRD7-IN-1, a modified derivative of BI7273 (BRD7/9 inhibitor), binds to a VHL ligand via a linker to form a PROTAC VZ185 (VZ185 against BRD7/9 with DC50s of 4.5 and 1.8 nM, respectively).

BRD7-IN-1, a modified derivative of BI7273 (BRD7/9 inhibitor), binds to a VHL ligand via a linker to form a PROTAC VZ185 (VZ185 against BRD7/9 with DC50s of 4.5 and 1.8 nM, respectively).

Specifications

Chemical Name or Material BRD7-IN-1
CAS 2448414-48-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H28Cl2N4O3
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (213.95 mM; Need ultrasonic) ∣DMSO : 30 mg/mL (64.19 mM; Need ultrasonic)
SMILES CN1C=C(C2=CC(OC)=C(CN3CCNCC3)C(OC)=C2)C(C=CN=C4)=C4C1=O.Cl.Cl
Molecular Weight (g/mol) 467.39
Formula Weight 467.39
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.