missing translation for 'onlineSavingsMsg'
Learn More

BRD4770, MedChemExpress

Product Code. 16447148
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16447148

Brand: Medchem Express HY16705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BRD4770 is a histone methyltransferase G9a inhibitor. BRD4770 reduces di- and trimethylation of lysine 9 on histone H3 (H3K9) with an EC50 of 5 μM, and has less or little effect toward H3K27me3, H3K36me3, H3K4me3, and H3K79me3. BRD4770 can activate the ataxia telangiectasia mutated (ATM) pathway and induce cell senescence.

BRD4770 is a histone methyltransferase G9a inhibitor. BRD4770 reduces di- and trimethylation of lysine 9 on histone H3 (H3K9) with an EC50 of 5 μM, and has less or little effect toward H3K27me3, H3K36me3, H3K4me3, and H3K79me3. BRD4770 can activate the ataxia telangiectasia mutated (ATM) pathway and induce cell senescence.

Specifications

Chemical Name or Material BRD4770
CAS 1374601-40-7
Color White
Purity Grade Notes Research
Molecular Formula C25H23N3O3
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (80.61 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2C(N=C(NC(C3=CC=CC=C3)=O)N2CCCC4=CC=CC=C4)=C1)OC
Molecular Weight (g/mol) 413.47
Formula Weight 413.47
Percent Purity 98.59%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.