Learn More
Invitrogen™ BRD4 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C131)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548328
Description
Specificity: This antibody recognizes BRD4 (Bromodomain-containing protein 4). It binds to a folded domain, amino acids 333-460. BRD4 is a chromatin reader protein that recognizes and binds acetylated histones and plays a key role in transmission of epigenetic memory.
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
Specifications
| BRD4 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| O60885 | |
| BRD4 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing BRD4 protein under non-denaturing conditions. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| RAB-C131 | |
| Unconjugated | |
| BRD4 | |
| Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23476 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.